Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FXYD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109745
Description
Immunogen sequence: MELVLVFLCS LLAPMVLASA AEKEKEMDPF HYDYQTLRIG GLVFAVVLFS VGILLILSRR CKCSFNQKPR APGDEEAQVE NLITANATEP QKAEN.
This gene encodes a member of the FXYD family of transmembrane proteins. This particular protein encodes phosphohippolin, which likely affects the activity of Na,K-ATPase. Multiple alternatively spliced transcript variants encoding the same protein have been described. Related pseudogenes have been identified on chromosomes 10 and X. Read-through transcripts have been observed between this locus and the downstream sodium/potassium-transporting ATPase subunit gamma (FXYD2, GeneID 486) locus.
Specifications
FXYD6 | |
Polyclonal | |
Unconjugated | |
FXYD6 | |
0610030I18Rik; FXYD domain containing ion transport regulator 6; FXYD domain-containing ion transport regulator 6; Fxyd6; Phosphohippolin; Php; PLM-like protein; Plp; UNQ521/PRO1056; Vascular endothelial cell-specific protein 6; VESP6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
59095, 63847 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.03 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q91XV6, Q9D164 | |
FXYD6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-95 of human FXYD6 (NP_071286.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction