Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FXYD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541057
Description
Peptide sequence: MATPTQTPTK APEEPDPFYY DYNTVQTVGM TLATILFLLG ILIVISKKVK Sequence homology: Cow: 77%; Human: 100%.
The specific function of this protein remains unknown.This reference sequence was derived from multiple replicate ESTs and validated by similar human genomic sequence. This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. FXYD2, also known as the gamma subunit of the Na, K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. This gene product, FXYD7, is novel and has not been characterized as a protein.
Specifications
| FXYD7 | |
| Polyclonal | |
| Unconjugated | |
| FXYD7 | |
| 1110035I01Rik; FLJ25096; FXYD domain containing ion transport regulator 7; FXYD domain-containing ion transport regulator 7; FXYD7; Unknown (protein for MGC:128487) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 53822 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P58549 | |
| FXYD7 | |
| Synthetic peptide directed towards the N-terminal of human FXYD7 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction