Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FYB/ADAP/SLAP130 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176810MFV450
Description
FYB/ADAP/SLAP130 Polyclonal antibody specifically detects FYB/ADAP/SLAP130 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
FYB/ADAP/SLAP130 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein | |
Antibody was raised against a 20 amino acid synthetic peptide from near the carboxy terminus of human ADAP. The immunogen is located within amino acids 640 - 690 of ADAP. Amino Acid Squence: LKGKDDRKKSIREKPKVSD | |
0.1 mL | |
Immunology | |
2533 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction