Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FYCO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104495 Supplier Invitrogen™ Supplier No. PA5104495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FYCO1.

This gene encodes a protein that contains a RUN domain, FYVE-type zinc finger domain and Golgi dynamics (GOLD) domain. The encoded protein plays a role in microtubule plus end-directed transport of autophagic vesicles through interactions with the small GTPase Rab7, phosphatidylinositol-3-phosphate (PI3P) and the autophagosome marker LC3. Mutations in this gene are a cause of autosomal recessive congenital cataract-2 (CATC2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FYCO1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FYCO1
Gene Accession No. Q9BQS8
Gene Alias 2810409M01Rik; CATC2; CTRCT18; DKFZp779K1152; FLJ13335; FYCO1; FYVE and coiled-coil domain autophagy adaptor 1; FYVE and coiled-coil domain containing 1; FYVE and coiled-coil domain-containing protein 1; maternal embryonic message 2; Mem2; MGC126517; MGC126519; RUFY3; RUN and FYVE domain containing 3; ZFYVE7; Zinc finger FYVE domain-containing protein 7
Gene Symbols FYCO1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FYCO1(Accession Q9BQS8), corresponding to amino acid residues E832-C882.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301085, 79443
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.