Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FYCO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113353
Description
FYCO1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a protein that contains a RUN domain, FYVE-type zinc finger domain and Golgi dynamics (GOLD) domain. The encoded protein plays a role in microtubule plus end-directed transport of autophagic vesicles through interactions with the small GTPase Rab7, phosphatidylinositol-3-phosphate (PI3P) and the autophagosome marker LC3. Mutations in this gene are a cause of autosomal recessive congenital cataract-2 (CATC2).
Specifications
| FYCO1 | |
| Polyclonal | |
| Unconjugated | |
| FYCO1 | |
| 2810409M01Rik; CATC2; CTRCT18; DKFZp779K1152; FLJ13335; FYCO1; FYVE and coiled-coil domain autophagy adaptor 1; FYVE and coiled-coil domain containing 1; FYVE and coiled-coil domain-containing protein 1; maternal embryonic message 2; Mem2; MGC126517; MGC126519; RUFY3; RUN and FYVE domain containing 3; ZFYVE7; Zinc finger FYVE domain-containing protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17281, 79443 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8VDC1, Q9BQS8 | |
| FYCO1 | |
| Recombinant Human FYVE and coiled-coil domain-containing protein 1 protein (2-255AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction