Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FYCO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119696 Supplier Invitrogen™ Supplier No. PA5119696
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: SK-Br-3 cell lysates, rat testis tissue, human liver tissue, human thyroid tissue, human colon tissue, mouse colon tissue, 293. Subcellular Location: Lysosome, Endosome, autophagosome.

This gene encodes a protein that contains a RUN domain, FYVE-type zinc finger domain and Golgi dynamics (GOLD) domain. The encoded protein plays a role in microtubule plus end-directed transport of autophagic vesicles through interactions with the small GTPase Rab7, phosphatidylinositol-3-phosphate (PI3P) and the autophagosome marker LC3. Mutations in this gene are a cause of autosomal recessive congenital cataract-2 (CATC2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FYCO1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene FYCO1
Gene Accession No. Q8VDC1, Q9BQS8
Gene Alias 2810409M01Rik; CATC2; CTRCT18; DKFZp779K1152; FLJ13335; FYCO1; FYVE and coiled-coil domain autophagy adaptor 1; FYVE and coiled-coil domain containing 1; FYVE and coiled-coil domain-containing protein 1; maternal embryonic message 2; Mem2; MGC126517; MGC126519; RUFY3; RUN and FYVE domain containing 3; ZFYVE7; Zinc finger FYVE domain-containing protein 7
Gene Symbols FYCO1
Host Species Rabbit
Immunogen Recombinant protein within human FYCO1 aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17281, 301085, 79443
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.