Learn More
Promega FYN A Kinase Enzyme System, 1mg
Catalog No. PRV3572 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3572 1 mg
1 options
Catalog No. PRV3572 Supplier Promega Supplier No. V3572
Add to Cart
Add to Cart
Request a Quote
Full-length recombinant human FYN A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FYN A (Fyn isoform A) is a member of the SRC tyrosine kinase oncogene family showing high homology to YES1, FGR and SRC.
  • Easily Screen and Profile FYN A Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C