Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fyn Antibody (SR2055), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Fyn |
---|---|
Clone | SR2055 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
Fyn Monoclonal antibody specifically detects Fyn in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Fyn | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.10, EC 2.7.10.2, FYN oncogene related to SRC, FGR, YES, MGC45350, OKT3-induced calcium influx regulator, p59-Fyn, protein-tyrosine kinase fyn, Proto-oncogene c-Fyn, Proto-oncogene Syn, proto-oncogene tyrosine-protein kinase fyn, SLKc-syn protooncogene, src/yes-related novel, Src-like kinase, SYN, tyrosine kinase p59fyn(T), tyrosine-protein kinase Fyn | |
A synthesized peptide derived from human Fyn (Uniprot #: P06241) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2055 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Phospho Specific, Protein Kinase, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2534 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title