Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FYN Mouse anti-Human, Mouse, Rat, Clone: 1G4B1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | FYN |
---|---|
Clone | 1G4B1 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7024
![]() ![]() |
Proteintech Group Inc
666061IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.Specifications
FYN | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FYN, p59 Fyn, Proto oncogene c Fyn, Proto oncogene Syn, SLK, Src like kinase, SYN, Tyrosine protein kinase Fyn | |
Fyn | |
IgG1 | |
Protein G | |
Antibody |
1G4B1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P06241, P39688, Q62844 | |
14360, 25150, 2534 | |
FYN Fusion Protein Ag11127 | |
Primary | |
-20°C | |
FYN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title