Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Fyn (p59 fyn) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS339R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS339R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS339R1 Supplier Epredia™ Supplier No. MS339R1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Fyn (p59fyn) Ab-1, Mouse Monoclonal Antibody.

p59fyn is a membrane-associated non-receptor protein tyrosine kinase that belongs to the Src-family of kinases. The unique N-terminal domain of p59fyn interacts with the CD3 and eta chains of the TcR. p59fyn can bind other proteins (p82 and p116) through its SH2 and SH3 domains, which may act as substrates or regulators of fyn activity. p59ffyn is highly expressed in brain suggesting that it may have a role in the sensory nervous network and in myelination at early stages of CNS formation.

Host Species: Mouse

Clone: 1S

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 26-75

Immunogen: Recombinant protein corresponding to aa 1-206 of murine p59fyn protein

Molecular Weight: 59 kDa

Positive Control: Jurkat cells, Lymphoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Kinase Assay (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Fyn (p59superscriptfynsuperscript) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1S
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Recombinant protein corresponding to aa 1-206 of murine p59superscriptfynsuperscriptf protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.