Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fyn Rabbit anti-Human, Mouse, Rat, Clone: 7Z9F2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
| Antigen | Fyn |
|---|---|
| Clone | 7Z9F2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Fyn Monoclonal antibody specifically detects Fyn in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Fyn | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2534 | |
| IgG | |
| Affinity purified |
| 7Z9F2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.10, EC 2.7.10.2, FYN oncogene related to SRC, FGR, YES, MGC45350, OKT3-induced calcium influx regulator, p59-Fyn, protein-tyrosine kinase fyn, Proto-oncogene c-Fyn, Proto-oncogene Syn, proto-oncogene tyrosine-protein kinase fyn, SLKc-syn protooncogene, src/yes-related novel, Src-like kinase, SYN, tyrosine kinase p59fyn(T), tyrosine-protein kinase Fyn | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Fyn (P06241). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title