Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FYTTD1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204090UV
Description
FYTTD1 Polyclonal antibody specifically detects FYTTD1 in Human, Mouse samples. It is validated for Western BlotSpecifications
FYTTD1 | |
Polyclonal | |
Western Blot | |
forty-two-three domain containing 1, Forty-two-three domain-containing protein 1, Protein 40-2-3, UAP56-interacting factor, UIFDKFZp761B1514 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human UAP56-Interacting Factor using the numbering given in entry NP_115664.2 (GeneID 84248). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
84248 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction