Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FYTTD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598705
Description
FYTTD1 Polyclonal Antibody for Western Blot, ELISA
Required for mRNA export from the nucleus to the cytoplasm. Acts as an adapter that uses the DDX39B/UAP56-NFX1 pathway to ensure efficient mRNA export and delivering to the nuclear pore. Associates with spliced and unspliced mRNAs simultaneously with ALYREF/THOC4. [UniProt].
Specifications
FYTTD1 | |
Polyclonal | |
Unconjugated | |
FYTTD1 | |
2010005M05Rik; 3830411L18Rik; 3830421F13Rik; 40-2-3 protein; 4933423A17Rik; Ac1176; AI098218; AW539567; C85190; forty-two-three domain containing 1; forty-two-three domain-containing protein 1; Forty-two-three domain-containing protein 1-like protein; FYTTD1; Protein 40-2-3; putative 40-2-3 protein; RGD1306899; UAP56-interacting factor; UAP56-interacting factor-like; UIF | |
Rabbit | |
Protein G | |
RUO | |
84248 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96QD9 | |
FYTTD1 | |
Recombinant Human UAP56-interacting factor protein (1-318AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction