Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FYTTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139990 Supplier Invitrogen™ Supplier No. PA5139990
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RKANLLRQNE GQRKPVAVLK RPSQLSRKNN IPANFTRSGN KLNHQKDTRQ ATFLFRRGLK VQAQLNTEQL LDDVVAKRTR QWR Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 90%.

Required for mRNA export from the nucleus to the cytoplasm. Acts as an adapter that uses the DDX39B/UAP56-NFX1 pathway to ensure efficient mRNA export and delivering to the nuclear pore. Associates with spliced and unspliced mRNAs simultaneously with ALYREF/THOC4. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FYTTD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FYTTD1
Gene Accession No. Q96QD9
Gene Alias 2010005M05Rik; 3830411L18Rik; 3830421F13Rik; 40-2-3 protein; 4933423A17Rik; Ac1176; AI098218; AW539567; C85190; forty-two-three domain containing 1; forty-two-three domain-containing protein 1; Forty-two-three domain-containing protein 1-like protein; FYTTD1; Protein 40-2-3; putative 40-2-3 protein; RGD1306899; UAP56-interacting factor; UAP56-interacting factor-like; UIF
Gene Symbols FYTTD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FYTTD1. Recombinant protein control fragment (Product #RP-109439).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84248
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.