Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FYTTD1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13238R

Description
FYTD1 required for mRNA export from the nucleus to the cytoplasm. This protein acts as an adapter that uses the BAT1/UAP56-NFX1 pathway to ensure efficient mRNA export and delivering to the nuclear pore. FYTD1 associates with spliced and unspliced mRNAs simultaneously with THOC4.Specifications
FYTTD1 | |
Polyclonal | |
Unconjugated | |
FYTTD1 | |
2010005M05Rik; 3830411L18Rik; 3830421F13Rik; 40-2-3 protein; 4933423A17Rik; Ac1176; AI098218; AW539567; BOS_828; C85190; forty-two-three domain containing 1; Forty-two-three domain-containing protein 1; Fyttd1; LOC100342818; Protein 40-2-3; RGD1306899; UAP56-interacting factor; UAP56-interacting factor-like; UIF; uncharacterized LOC100342818 | |
Rabbit | |
Protein A | |
RUO | |
84248 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96QD9 | |
FYTTD1 | |
KLH conjugated synthetic peptide derived from human FYTTD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction