Learn More
Invitrogen™ FZD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595287
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: 22RV1 whole cell, 293T whole cell. IHC: human prostatic cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD1 protein contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, 7 transmembrane domains, and a C-terminal PDZ domain-binding motif. The FZD1 transcript is expressed in various tissues.
Specifications
FZD1 | |
Polyclonal | |
Unconjugated | |
Fzd1 | |
AW227548; Chfz-1; Dfrizzled-1; DKFZp564G072; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 1; FLJ95923; frizzled; frizzled 1; frizzled 1, seven transmembrane spanning receptor; frizzled class receptor 1; frizzled family receptor 1; frizzled homolog 1; frizzled homolog 1 (Drosophila); frizzled, Drosophila, homolog of, 1; frizzled-1; Fz receptor 1; Fz1; FZ-1; Fzd1; FzE1; hFz1; mFz1; rFz1; Wnt receptor; Xfrizzled-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8321 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q9UP38 | |
Fzd1 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human Frizzled homolog 1 (369-400aa YIAGFLLEDRVVCNDKFAEDGARTVAQG TKKE). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.