Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115257
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115257 Supplier Thermo Scientific Supplier No. PA5115257
Add to Cart
Add to Cart

FZD1 Polyclonal Antibody

FZD1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Fzd1
Gene Accession No. Q08463, Q9UP38
Gene Alias AW227548; Chfz-1; Dfrizzled-1; DKFZp564G072; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 1; FLJ95923; frizzled; frizzled 1; frizzled 1, seven transmembrane spanning receptor; frizzled class receptor 1; frizzled family receptor 1; frizzled homolog 1; frizzled homolog 1 (Drosophila); frizzled, Drosophila, homolog of, 1; frizzled-1; Fz receptor 1; Fz1; FZ-1; Fzd1; FzE1; hFz1; mFz1; rFz1; Wnt receptor; Xfrizzled-1
Gene Symbols Fzd1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FZD1(Accession Q9UP38), corresponding to amino acid residues L21-R71.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58868, 8321
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.