Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547072
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547072 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547072 Supplier Invitrogen™ Supplier No. PA547072
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 10% cross-reactivity with recombinant mouse (rm) Frizzled-2, rmFrizzled-4, rmFrizzled-7, and rmFrizzled-8 is observed, and less than 1% cross-reactivity with rmFrizzled-3 and rmFrizzled-6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD1 protein contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, 7 transmembrane domains, and a C-terminal PDZ domain-binding motif. The FZD1 transcript is expressed in various tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Fzd1
Gene Accession No. O70421
Gene Alias AW227548; Chfz-1; Dfrizzled-1; DKFZp564G072; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 1; FLJ95923; frizzled; frizzled 1; frizzled 1, seven transmembrane spanning receptor; frizzled class receptor 1; frizzled family receptor 1; frizzled homolog 1; frizzled homolog 1 (Drosophila); frizzled, Drosophila, homolog of, 1; frizzled-1; Fz receptor 1; Fz1; FZ-1; Fzd1; FzE1; hFz1; mFz1; rFz1; Wnt receptor; Xfrizzled-1
Gene Symbols Fzd1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-1 Gln72-His248 (Met122Ile).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14362
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.