Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FZD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA533550
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Panda, Dog, Bat, Bovine (100%).
Members of the ′frizzled′ gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD1 protein contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, 7 transmembrane domains, and a C-terminal PDZ domain-binding motif. The FZD1 transcript is expressed in various tissues.
Specifications
| FZD1 | |
| Polyclonal | |
| Unconjugated | |
| FZD1 | |
| FZD1, Chfz-1, Fz1, Frizzled 1, Fz receptor 1, HFz1, Frizzled homolog 1, Fz-1, FzE1, Rfz1, Xfrizzled-1, Wnt receptor, Dfrizzled-1, Frizzled family receptor 1, Frizzled-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8321 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9UP38 | |
| FZD1 | |
| Synthetic 13 amino acid peptide from N-terminal extracellular domain of human FZD1 / Frizzled 1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction