Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130050
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130050 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130050 Supplier Invitrogen™ Supplier No. PA130050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-30050 detects Frizzled from mouse samples. The immunogen shares 91% sequence homology with Rat.

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD1 protein contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, 7 transmembrane domains, and a C-terminal PDZ domain-binding motif. The FZD1 transcript is expressed in various tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Fzd1
Gene Accession No. O70421
Gene Alias AW227548; Chfz-1; Dfrizzled-1; DKFZp564G072; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 1; FLJ95923; frizzled; frizzled 1; frizzled 1, seven transmembrane spanning receptor; frizzled class receptor 1; frizzled family receptor 1; frizzled homolog 1; frizzled homolog 1 (Drosophila); frizzled, Drosophila, homolog of, 1; frizzled-1; Fz receptor 1; Fz1; FZ-1; Fzd1; FzE1; hFz1; mFz1; rFz1; Wnt receptor; Xfrizzled-1
Gene Symbols Fzd1
Host Species Rabbit
Immunogen Mouse Fz7 residues 1-12 (MRGPGTAASHSPC).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14362
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.