Learn More
Invitrogen™ FZD10 Polyclonal Antibody

Description
Sequence homology: Rat (88%).
Specifications
Specifications
| Antigen | FZD10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 50% glycerol and 0.025% sodium azide |
| Gene | FZD10 |
| Gene Accession No. | Q8BKG4, Q9ULW2 |
| Gene Alias | CD350; CD350 antigen; frizzled 10, seven transmembrane spanning receptor; frizzled class receptor 10; frizzled family receptor 10; frizzled homolog 10; frizzled homolog 10 (Drosophila); frizzled homolog 10, pseudogene 1; frizzled-10; Fz10; Fz-10; Fzd10; Fzd10-ps1; FzE7; hFz10 |
| Gene Symbols | FZD10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.