Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533554
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533554 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533554 Supplier Invitrogen™ Supplier No. PA533554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FZD3 Polyclonal Antibody for IHC (P)

This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene FZD3
Gene Accession No. Q9NPG1
Gene Alias AU020229; D930050A07Rik; frizzled 3; frizzled 3, seven transmembrane spanning receptor; frizzled class receptor 3; frizzled family receptor 3; frizzled homolog 3; frizzled homolog 3 (Drosophila); frizzled-3; Fz3; Fz-3; FZD3; hFz3; mFz3
Gene Symbols FZD3
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human FZD3 / Frizzled 3.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7976
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.