Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533555
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533555 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533555 Supplier Invitrogen™ Supplier No. PA533555
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Rat, Dolphin, Hamster, Elephant, Panda, Dog, Bovine (100%).

FZD4 is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. Frizzled 4 is widely expressed, particularly in heart, skeletal muscle, ovary, and fetal kidney. Transcripts have also been identified in liver, kidney, pancreas, spleen, and fetal lung, and in smaller amounts in placenta, adult lung, prostate, testis, colon, and fetal brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene FZD4
Gene Accession No. Q9QZH0, Q9ULV1
Gene Alias CD344; CD344 antigen; EVR1; FEVR; frizzled 4, seven transmembrane spanning receptor; frizzled class receptor 4; frizzled family receptor 4; frizzled homolog 4; frizzled homolog 4 (Drosophila); frizzled receptor 4; frizzled-4; Fz4; Fz-4; Fzd4; FZD4S; FzE4; GPCR; hFz4; mFz4; MGC34390; rFz4; WNT receptor frizzled-4
Gene Symbols FZD4
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from N-terminal extracellular domain of human FZD4 / Frizzled 4.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100756296, 445416, 485149, 64558, 8322
Target Species Human, Rat, Canine, Monkey, Bovine, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.