Learn More
Invitrogen™ FZD5 Monoclonal Antibody (2D12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517080
Description
MA5-17080 targets FZD5 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17080 immunogen is purified recombinant fragment of human FZD5 (amino acids:151-217) expressed in E. Coli. MA5-17080 detects FZD5 which has a predicted molecular weight of approximately 64.5kDa.
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD5 protein is believed to be the receptor for the Wnt5A ligand.
Specifications
FZD5 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q13467 | |
FZD5 | |
Purified recombinant fragment of human FZD5 (amino acids:151-217) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
2D12 | |
Unconjugated | |
FZD5 | |
C2orf31; DKFZp434E2135; frizzled 5, seven transmembrane spanning receptor; frizzled class receptor 5; frizzled family receptor 5; frizzled-5; Fz5; Fz-5; Fzd5; Fzd-5; FzE5; HFZ5; MGC129692; seven-transmembrane receptor frizzled-5; Wnt receptor | |
Mouse | |
Protein G | |
RUO | |
7855 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.