Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533557
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533557 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533557 Supplier Invitrogen™ Supplier No. PA533557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Mouse, Rat, Hamster, Elephant (100%) Dog, Rabbit (94%).

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD5 protein is believed to be the receptor for the Wnt5A ligand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene FZD5
Gene Accession No. Q13467, Q8CHL0, Q9EQD0
Gene Alias C2orf31; DKFZp434E2135; frizzled 5, seven transmembrane spanning receptor; frizzled class receptor 5; frizzled family receptor 5; frizzled-5; Fz5; Fz-5; Fzd5; Fzd-5; FzE5; HFZ5; MGC129692; seven-transmembrane receptor frizzled-5; Wnt receptor
Gene Symbols FZD5
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminal extracellular domain of human FZD5 / Frizzled 5.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100762351, 14367, 317674, 7855
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.