Learn More
Invitrogen™ FZD5 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey, Mouse, Rat, Elephant, Dog, Bovine, Rabbit, Pig (94%).
Specifications
Specifications
| Antigen | FZD5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | FZD5 |
| Gene Accession No. | Q13467 |
| Gene Alias | C2orf31; DKFZp434E2135; frizzled 5, seven transmembrane spanning receptor; frizzled class receptor 5; frizzled family receptor 5; frizzled-5; Fz5; Fz-5; Fzd5; Fzd-5; FzE5; HFZ5; MGC129692; seven-transmembrane receptor frizzled-5; Wnt receptor |
| Gene Symbols | FZD5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.