Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588750 Supplier Invitrogen™ Supplier No. PA588750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RMSCDRLPVL GRDAEVLCMD YNRSEATTAP PRPFPAKPTL PGPPGAPASG GECPAGGPFV CKCREPFVPI LKESHPLYNK VRTGQVPNCA VPCYQPSFSA DERTFATFWI G; Positive Samples: Rat kidney; Cellular Location: Cell membrane, Golgi apparatus membrane, Multi-pass membrane protein.

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD5 protein is believed to be the receptor for the Wnt5A ligand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FZD5
Gene Accession No. Q13467, Q8CHL0
Gene Alias C2orf31; DKFZp434E2135; frizzled 5, seven transmembrane spanning receptor; frizzled class receptor 5; frizzled family receptor 5; frizzled-5; Fz5; Fz-5; Fzd5; Fzd-5; FzE5; HFZ5; MGC129692; seven-transmembrane receptor frizzled-5; Wnt receptor
Gene Symbols FZD5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 130-240 of human FZD5 (NP_0034592).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317674, 7855
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.