Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587953 Supplier Invitrogen™ Supplier No. PA587953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSLFTCEPIT VPRCMKMAYN MTFFPNLMGH YDQSIAAVEM EHFLPLANLE CSPNIETFLC KAFVPTCIEQ IHVVPPCRKL CEKVYSDCKK LIDTFGIRWP EELECDRLQY CDETVPVTFD PHTEFLGPQK KTEQVQRDIG FWCPRHLKTS GGQGYKFLGI DQCAPPCPNM YFKSDELEFA KSF; Positive Samples: mouse kidney, rat kidney; Cellular Location: Apical cell membrane, Cell membrane, Cell surface, Cytoplasmic vesicle membrane, Membrane, Multi-pass membrane protein.

The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FZD6
Gene Accession No. Q61089
Gene Alias frizzled 6; frizzled 6, seven transmembrane spanning receptor; frizzled class receptor 6; frizzled family receptor 6; frizzled homolog 6; frizzled homolog 6 (Drosophila); Frizzled6; frizzled-6; FZ6; Fz-6; Fzd6; FZD-6; HFZ6; mFz6; NDNC10; seven transmembrane helix receptor
Gene Symbols FZD6
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14368, 282581
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.