Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FZD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575511
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human.
Specifications
FZD6 | |
Polyclonal | |
Unconjugated | |
FZD6 | |
frizzled 6; frizzled 6, seven transmembrane spanning receptor; frizzled class receptor 6; frizzled family receptor 6; frizzled homolog 6; frizzled homolog 6 (Drosophila); Frizzled6; frizzled-6; FZ6; Fz-6; Fzd6; FZD-6; HFZ6; mFz6; NDNC10; seven transmembrane helix receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14368, 282581, 8323 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O60353, Q61089 | |
FZD6 | |
Synthetic peptide corresponding to amino acids 169-214 of Human FZD6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction