Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533559
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533559 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533559 Supplier Invitrogen™ Supplier No. PA533559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FZD7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD7 protein contains an N-terminal signal sequence, 10 cysteine residues typical of the cysteine-rich extracellular domain of Fz family members, 7 putative transmembrane domains, and an intracellular C-terminal tail with a PDZ domain-binding motif. FZD7 gene expression may downregulate APC function and enhance beta-catenin-mediated signals in poorly differentiated human esophageal carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene FZD7
Gene Accession No. O75084, Q61090
Gene Alias frizzled 7, seven transmembrane spanning receptor; frizzled class receptor 7; frizzled family receptor 7; frizzled homolog 7; frizzled homolog 7 (Drosophila); Frizzled, drosophila, homolog of, 7; frizzled-7; Fz7; Fz-7; FZD7; FzE3; hFz7; LOC100360552; mFz7; rCG22430-like; rizzled 7
Gene Symbols FZD7
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from N-terminal extracellular domain of human FZD7 / Frizzled 7.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360552, 100513691, 100766658, 103348917, 14369, 488478, 524255, 8324
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.