Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111642 Supplier Invitrogen™ Supplier No. PA5111642
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVGQNTSDGS GGPGGGPTAY PTAPYLPDLP FTAL Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 88%.

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD7 protein contains an N-terminal signal sequence, 10 cysteine residues typical of the cysteine-rich extracellular domain of Fz family members, 7 putative transmembrane domains, and an intracellular C-terminal tail with a PDZ domain-binding motif. FZD7 gene expression may downregulate APC function and enhance beta-catenin-mediated signals in poorly differentiated human esophageal carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FZD7
Gene Accession No. O75084
Gene Alias frizzled 7, seven transmembrane spanning receptor; frizzled class receptor 7; frizzled family receptor 7; frizzled homolog 7; frizzled homolog 7 (Drosophila); Frizzled, drosophila, homolog of, 7; frizzled-7; Fz7; Fz-7; FZD7; FzE3; hFz7; LOC100360552; mFz7; rCG22430-like; rizzled 7
Gene Symbols FZD7
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human FZD7. Recombinant protein control fragment (Product #RP-107678).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8324
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.