Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FZD8 Polyclonal Antibody
FZD8 Polyclonal Antibody
Supplier: Thermo Scientific PA550589
Description
FZD8 Polyclonal Antibody for IHC (P)

Specifications
FZD8 | |
Polyclonal | |
Unconjugated | |
Fzd8 | |
frizzled 8, seven transmembrane spanning receptor; frizzled class receptor 8; frizzled family receptor 8; frizzled homolog 8; frizzled homolog 8 (Drosophila); frizzled-8; Fz8; Fz-8; Fzd8; hFZ8; HGNC:4046 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14370, 364754, 8325 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q498S8, Q61091, Q9H461 | |
Fzd8 | |
Synthetic peptide corresponding to residues near the C terminal of human frizzled family receptor 8. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction