Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111669 Supplier Invitrogen™ Supplier No. PA5111669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QCRAPMVSVS SERHPLYNRV KTGQIANCA Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

Members of the Frizzled family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. Frizzled 8 is highly expressed in two human cancer cell lines, indicating that it may play a role in several types of cancer. Its expression has been reported in brain, heart, kidney, pancreas, and skeletal muscle. ESTs have been isolated from embryo, eye, head/neck, heart, kidney, liver/spleen, pancreas, prostate, and testis libraries, as well as many tumor libraries. The crystal structure of the extracellular cysteine-rich domain of a similar mouse protein has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fzd8
Gene Accession No. Q9H461
Gene Alias frizzled 8, seven transmembrane spanning receptor; frizzled class receptor 8; frizzled family receptor 8; frizzled homolog 8; frizzled homolog 8 (Drosophila); frizzled-8; Fz8; Fz-8; Fzd8; hFZ8; HGNC:4046
Gene Symbols Fzd8
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human FZD8. Recombinant protein control fragment (Product #RP-107796).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8325
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.