Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FZD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120635
This item is not returnable.
View return policy
Description
Positive test controls include: 293T, A-549, HeLa, Mouse liver, Rat liver. Immunogen sequence: PDRMRCDRLP EQGNPDTLCM DYNRTDLTTA APSPPRRLPP PPPGEQPPSG SGHGRPPGAR PPHRGGGRGG GGGDAAAPPA RGGGGGGKAR PPGGGAAPCE PGCQCRAPMV SVSSERHPLY NRVKTGQIAN CALPCHNPFF SQDERA.
Members of the Frizzled family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. Frizzled 8 is highly expressed in two human cancer cell lines, indicating that it may play a role in several types of cancer. Its expression has been reported in brain, heart, kidney, pancreas, and skeletal muscle. ESTs have been isolated from embryo, eye, head/neck, heart, kidney, liver/spleen, pancreas, prostate, and testis libraries, as well as many tumor libraries. The crystal structure of the extracellular cysteine-rich domain of a similar mouse protein has been determined.
Specifications
| FZD8 | |
| Polyclonal | |
| Unconjugated | |
| Fzd8 | |
| frizzled 8, seven transmembrane spanning receptor; frizzled class receptor 8; frizzled family receptor 8; frizzled homolog 8; frizzled homolog 8 (Drosophila); frizzled-8; Fz8; Fz-8; Fzd8; hFZ8; HGNC:4046 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14370, 364754, 8325 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.96 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q498S8, Q61091, Q9H461 | |
| Fzd8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-275 of human FZD8 (NP_114072.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction