Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FZR1/CDH1 Antibody (AR38.2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP219018AF700
Description
FZR1/CDH1 Monoclonal specifically detects FZR1/CDH1 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
FZR1/CDH1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FZR1 | |
His-tagged recombinant FZR1/CDH1 | |
0.1 mL | |
Cell Cycle and Replication | |
51343 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
AR38.2 | |
Western Blot, ELISA | |
CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242fizzy-related protein homolog | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction