Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZR1 Monoclonal Antibody (1621CT637.76.67)
GREENER_CHOICE

Catalog No. PIMA537699
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537699 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537699 Supplier Invitrogen™ Supplier No. MA537699
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FZR1 Monoclonal Antibody for Western Blot

FZR is a 7WD domain family member implicated in cell cycle regulation of Drosophila and yeast. It has been shown that the over expression of fzr proteins increases natural killer cell-mediated cell death and as a result, suppresses tumor growth. The fzr protein is essential for cyclosome activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZR1
Applications Western Blot
Classification Monoclonal
Clone 1621CT637.76.67
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FZR1
Gene Accession No. Q9UM11
Gene Alias AW108046; CDC20 homolog 1; CDC20C; CDC20-like 1b; CDC20-like protein 1; Cdh1; Cdh1/Hct1 homolog; cell division cycle 20 related 1; fizzy/cell division cycle 20 related 1; fizzy/cell division cycle 20 related 1 (Drosophila); fizzy-related protein homolog; Fyr; FZR; Fzr1; FZR2; HCDH; HCDH1; KIAA1242
Gene Symbols FZR1
Host Species Mouse
Immunogen recombinant protein between1-496 amino acids from human FZR.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.