Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FZR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100535 Supplier Invitrogen™ Supplier No. PA5100535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FZR1.

FZR is a 7WD domain family member implicated in cell cycle regulation of Drosophila and yeast. It has been shown that the over expression of fzr proteins increases natural killer cell-mediated cell death and as a result, suppresses tumor growth. The fzr protein is essential for cyclosome activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FZR1
Gene Accession No. Q9R1K5, Q9UM11
Gene Alias AW108046; CDC20 homolog 1; CDC20C; CDC20-like 1b; CDC20-like protein 1; Cdh1; Cdh1/Hct1 homolog; cell division cycle 20 related 1; fizzy/cell division cycle 20 related 1; fizzy/cell division cycle 20 related 1 (Drosophila); fizzy-related protein homolog; Fyr; FZR; Fzr1; FZR2; HCDH; HCDH1; KIAA1242
Gene Symbols FZR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FZR1(Accession Q9UM11), corresponding to amino acid residues Q118-T168.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314642, 51343, 56371
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.