Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-CSF Antibody (CSF3/900), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247934MFV450
Description
G-CSF Monoclonal antibody specifically detects G-CSF in Human, Macaque samples. It is validated for Flow CytometrySpecifications
G-CSF | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Macaque | |
Purified |
Flow Cytometry | |
CSF3/900 | |
Flow Cytometry | |
C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
Recombinant full-length human G-CSF protein (Uniprot: P09919) | |
0.1 mL | |
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research | |
1440 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction