Learn More
Invitrogen™ G-CSF Polyclonal Antibody

Description
Neutralization:To yield one-half maximal inhibition (ND50) of the biological activity of mG-CSF (0.5 ng/mL), a 0.005-0.010 μg/mL of this antibody is required. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | G-CSF |
| Applications | ELISA, Neutralization, Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Csf3 |
| Gene Accession No. | P09920 |
| Gene Alias | C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF 3; CSF3; CSF3OS; Csfg; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; granulocyte-colony stimulating factor; gsf3; lenograstim; MGC45931; MGI-IG; Pluripoietin; RP23-395E10.4 |
| Gene Symbols | Csf3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.