Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech G-CSF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 500P69BT50
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P69BT50 50 μg
500P69BT25 25 μg
50-271-8458 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P69BT50 Supplier PeproTech Supplier No. 500P69BT50UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine G-CSF. Anti-Murine G-CSF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mG-CSF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine G-CSF (500-P69) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mG-CSF. Western Blot: To detect mG-CSF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mG-CSF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P69BT-1MG will be provided as 2 x 500 μg

G-CSF (Granulocyte colony stimulating factor) is a naturally occurring cytokine that stimulates the production and antibacterial function of neutrophils and monocytes. Human G-CSF is an 18.8 kDa protein containing 175 amino acid residues, and a soluble isoform of the G-CSF receptor has been described. The pleotropic cytokine is produced by activated monocytes, macrophages, endothelial cells, fibroblasts, astrocytes, osteoblasts and bone marrow cells. G-CSF has been shown to have specific effects on the proliferation, differentiation and activation of hematopoietic cells. G-CSF is also expressed by various transformed cells such as carcinoma cells and myeloblastic leukemia cells. G-CSF is encoded by two distinct DNA sequences, resulting in a full size, high activity and a shorter, low activity isoform of G-CSF. G-CSF is highly conserved among species and has been shown to exert its biological functions through interaction with its receptor expressed on the surface of hematopoietic progenitors, neutrophilic granulocytes and certain carcinoma cell lines. Clinical use of G-CSF has been approved for several therapeutic applications, treatment of neonatal infections, therapy of acute myocardial infarction, granulocyte transfusion in patients with neutropenia, in severe infections and sepsis, therapy in chronic autoimmune neutropenia, treatment of acute myeloid leukemias, Sweet's syndrome and AIDS. Further, G-CSF has been shown to be a marker protein for different carcinomas such as bladder cancer and dysfunction of the protein has been linked to Kostmann Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G-CSF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Csf3
Gene Accession No. P09920
Gene Alias C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF 3; CSF3; CSF3OS; Csfg; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; granulocyte-colony stimulating factor; gsf3; lenograstim; MGC45931; MGI-IG; Pluripoietin; RP23-395E10.4
Gene Symbols Csf3
Host Species Rabbit
Immunogen E.coli-derived Recombinant Murine G-CSF.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12985
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.