Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-CSFR/CD114 Antibody (38660), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB381Y
This item is not returnable.
View return policy
Description
G-CSFR/CD114 Monoclonal antibody specifically detects G-CSFR/CD114 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
G-CSFR/CD114 | |
Monoclonal | |
DyLight 680 | |
CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
Mouse myeloma cell line NS0-derived recombinant human G-CSF R/CD114, Glu25-Pro621, Accession # Q99062 | |
0.5 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
38660 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1441 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction