Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-CSFR/CD114 Antibody (723806) [Alexa Fluor™ 532], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB60391X
Description
G-CSFR/CD114 Monoclonal antibody specifically detects G-CSFR/CD114 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
G-CSFR/CD114 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1441 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
723806 | |
Flow Cytometry, CyTOF-ready | |
CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
Mouse myeloma cell line NS0-derived recombinant mouse G-CSF R/CD114, Cys26-Asp626, Accession # P40223 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction