Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G gamma12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982733
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2733 Supplier Bioss Supplier No. BS13242R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. G proteins are composed of 3 units, alpha, beta and gamma.The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.

Specifications

Antigen G gamma12
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GNG12
Gene Accession No. Q9UBI6
Gene Alias 2010305F15Rik; AA536815; AI115529; AI314170; AI842738; G protein subunit gamma 12; GNG12; G-protein gamma-12 subunit; guanine nucleotide binding protein (G protein), gamma 12; Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-12
Gene Symbols GNG12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human G gamma12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55970
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.