Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G gamma14 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | G gamma14 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
G gamma14 Polyclonal specifically detects G gamma14 in Human, Mouse samples. It is validated for Western Blot.Specifications
G gamma14 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
G protein cone gamma 8 subunit, gamma-T2 subunit, G-GAMMA-8, G-gamma-9, G-GAMMA-C, GNG8, GNG9G gamma-C, GNGT8, guanine nucleotide binding protein (G protein), gamma transducing activitypolypeptide 2, guanine nucleotide binding protein gamma 9, Guanine nucleotide binding protein gamma transducing activity polypeptide 2, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-T2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-69 of human G gamma14 (NP_113686.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS with 50% glycerol, pH7.3. | |
2793 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title