Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G gamma7 Antibody (OTI3G1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270838AF350
Description
G gamma7 Monoclonal specifically detects G gamma7 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
G gamma7 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
GNG7 | |
Recombinant protein fragment corresponding to amino acids 2-65 of human G gamma7 (NP_443079) produced in E.coli. | |
0.1 mL | |
2788.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3G1 | |
Western Blot | |
FLJ00058, GNGT7, guanine nucleotide binding protein (G protein), gamma 7, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-7 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction