Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alpha 12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985904
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5904 Supplier Bioss Supplier No. BS4109R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Guanine nucleotide-binding proteins (G proteins) are modulators or transducers in various transmembrane signaling systems. G proteins are composed of 3 subunits; alpha, beta and gamma. The alpha chain contains the guanine nucleotide binding site.

Specifications

Antigen G protein alpha 12
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GNA12
Gene Accession No. Q03113
Gene Alias AI414047; AI504261; G alpha-12; G protein subunit alpha 12; Galpha12; gep; Gna12; Gna-12; G-protein subunit alpha-12; guanine nucleotide binding protein (G protein) alpha 12; guanine nucleotide binding protein, alpha 12; Guanine nucleotide-binding protein subunit alpha-12; NNX3; RMP; WUGSC:H_GS165O14.2
Gene Symbols GNA12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GNA12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2768
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.