Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G protein alpha 16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30347220UL
Description
G protein alpha 16 Polyclonal specifically detects G protein alpha 16 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
G protein alpha 16 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
G alpha-15, G alpha-16, GNA16G-protein subunit alpha-15, G-protein subunit alpha-16, guanine nucleotide binding protein (G protein), alpha 15 (Gq class), guanine nucleotide-binding protein subunit alpha-15, Guanine nucleotide-binding protein subunit alpha-16 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-374 of human G protein alpha 16 (NP_002059.3). | |
20 μL | |
Signal Transduction | |
2769 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction