Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alpha inhibitor 1 (GNAI1) Monoclonal Antibody (OTI2D1), TrueMAB™, OriGene
SDP

Catalog No. 502543894
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3894 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3894 Supplier OriGene Supplier No. CF811907
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

G protein alpha inhibitor 1 (GNAI1) Monoclonal antibody specifically detects G protein alpha inhibitor 1 (GNAI1) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen G protein alpha inhibitor 1 (GNAI1)
Applications Western Blot
Classification Monoclonal
Clone OTI2D1
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Uba3
Gene Accession No. Q8C878, Q8TBC4, Q99MI7
Gene Alias adenylate cyclase-inhibiting G alpha protein; AU046200; BPGTPB; G protein subunit alpha i1; Gi; Gi1 protein alpha subunit; Gi1 protein alpha-subunit; Gialpha1; GNAI1; Gnai-1; GNAI1 protein; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; guanine nucleotide binding protein, alpha inhibiting 1; guanine nucleotide-binding protein G(i) subunit alpha-1; guanine nucleotide-binding protein G(i), alpha-1 subunit; p-alpha-h subunit; unnamed protein product
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-463 of human UBA3 (NP_003959) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117553, 22200, 9039
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.