Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alpha inhibitor 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982740
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2740 Supplier Bioss Supplier No. BS13249R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Guanine nucleotide-binding proteins (G proteins) form a large family of signal-transducing molecules. They are found as heterotrimers made up of alpha, beta, and gamma subunits. Members of the G protein family have been characterized most extensively on the basis of the alpha subunit, which binds guanine nucleotide, is capable of hydrolyzing GTP, and interacts with specific receptor and effector molecules. The G protein family includes Gs and Gi, the stimulatory and inhibitory GTP-binding regulators of adenylate cyclase; Go, a protein abundant in brain; and transducin-1 and transducin-2, proteins involved in phototransduction in retinal rods and cones, respectively.

Specifications

Antigen G protein alpha inhibitor 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GNAI1
Gene Accession No. P63096
Gene Alias Adenylate cyclase-inhibiting G alpha protein; AU046200; BOS_4157; BPGTPB; G protein subunit alpha i1; Gi; Gi1 protein alpha subunit; Gi1 protein alpha-subunit; Gialpha1; Gnai1; Gnai-1; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; guanine nucleotide binding protein, alpha inhibiting 1; Guanine nucleotide-binding protein G(i) subunit alpha-1; guanine nucleotide-binding protein G(i), alpha-1 subunit
Gene Symbols GNAI1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human G protein alpha inhibitor 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2770
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.