Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G Protein alpha z Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | G Protein alpha z |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
G Protein alpha z Polyclonal specifically detects G Protein alpha z in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
G Protein alpha z | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
2781 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
G(x) alpha chain, guanine nucleotide binding protein (G protein), alpha z polypeptide, guanine nucleotide-binding protein G(z) subunit alpha, gz-alpha, transducin alpha | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human G Protein alpha z (NP_002064.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title